Product References
Bcl-xL mutant promotes cartilage differentiation of BMSCs by upregulating TGF-β/BMP expression levels.
Experimental and therapeutic medicine
Xiao K,Yang L,Xie W,Gao X,Huang R,Xie M
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 12-0903-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Adipose-derived stem cells therapy effectively attenuates PM2.5-induced lung injury.
Stem cell research & therapy
Gao J,Yuan J,Liu Q,Wang Y,Wang H,Chen Y,Ding W,Ji G,Lu Z
12-0903-82 was used in Flow Cytometry to suggest that ADSC transplantation may be a potential therapeutic approach for severe air pollution-associated diseases.
Sat Jun 19 00:00:00 EDT 2021
Mechanisms of interleukin 4 mediated increase in efficacy of vaccines against opioid use disorders.
NPJ vaccines
Crouse B,Robinson C,Huseby Kelcher A,Laudenbach M,Abrahante JE,Pravetoni M
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 12-0903-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
Nestin-GFP transgene labels skeletal progenitors in the periosteum.
Bone
Tournaire G,Stegen S,Giacomini G,Stockmans I,Moermans K,Carmeliet G,van Gastel N
12-0903-82 was used in Flow Cytometry to show that in adult mice, Nes-GFP cells are present in the periosteum and localize closely to blood vessels, but periosteal Nes-GFP cells express SSC and progenitor markers differently compared to Nes-GFP cells in the bone marrow.
Wed Apr 01 00:00:00 EDT 2020
Low-Dose Radiotherapy Ameliorates Advanced Arthritis in hTNF-α tg Mice by Particularly Positively Impacting on Bone Metabolism.
Frontiers in immunology
Deloch L,Derer A,Hueber AJ,Herrmann M,Schett GA,Wölfelschneider J,Hahn J,Rühle PF,Stillkrieg W,Fuchs J,Fietkau R,Frey B,Gaipl US
12-0903 was used in Flow cytometry/Cell sorting to study the underlying mechanisms by which low-dose radiotherapy mediates anti-inflammatory and pain-reducing effects in non-linear dose response relationships.
Mon Sep 30 00:00:00 EDT 2019
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
12-0903 was used in Flow cytometry/Cell sorting to indicate that Rap1 is essential to maintain the immunomodulatory function of MSCs.
Wed Mar 07 00:00:00 EST 2018
Low-Dose Radiotherapy Has No Harmful Effects on Key Cells of Healthy Non-Inflamed Joints.
International journal of molecular sciences
Deloch L,Rückert M,Fietkau R,Frey B,Gaipl US
12-0903 was used in Flow cytometry/Cell sorting to show that low-dose radiotherapy with particularly a single dose of 0.5 Gray has no harmful effects on cells of healthy joints.
Tue Oct 16 00:00:00 EDT 2018
Differential regulation of C5a receptor 1 in innate immune cells during the allergic asthma effector phase.
PloS one
Ender F,Wiese AV,Schmudde I,Sun J,Vollbrandt T,König P,Laumonnier Y,Köhl J
12-0903 was used in Flow cytometry/Cell sorting to demonstrate a complex regulation pattern of C5aR1 in the airways, lung tissue and lymph nodes of mice.
Thu Aug 17 00:00:00 EDT 2017
Mononuclear Phagocytes Are Dispensable for Cardiac Remodeling in Established Pressure-Overload Heart Failure.
PloS one
Patel B,Ismahil MA,Hamid T,Bansal SS,Prabhu SD
12-0903 was used in Flow cytometry/Cell sorting to assess for alterations in MPs.
Thu Aug 10 00:00:00 EDT 2017
A New Method to Stabilize C-Kit Expression in Reparative Cardiac Mesenchymal Cells.
Frontiers in cell and developmental biology
Wysoczynski M,Dassanayaka S,Zafir A,Ghafghazi S,Long BW,Noble C,DeMartino AM,Brittian KR,Bolli R,Jones SP
12-0903 was used in Flow cytometry/Cell sorting to investigate a new method for c-kit expressing cell isolation and stabilisation in reparative cardiac mesenchymal cells.
Thu Aug 18 00:00:00 EDT 2016
Multi-kilobase homozygous targeted gene replacement in human induced pluripotent stem cells.
Nucleic acids research
Byrne SM,Ortiz L,Mali P,Aach J,Church GM
12-0903 was used in Flow cytometry/Cell sorting to assess the efficiency of homologous recombination after trialling different targeting vector design parameters.
Wed Feb 18 00:00:00 EST 2015
Collaborative rewiring of the pluripotency network by chromatin and signalling modulating pathways.
Nature communications
Tran KA,Jackson SA,Olufs ZP,Zaidan NZ,Leng N,Kendziorski C,Roy S,Sridharan R
Published figure using CD90.2 (Thy-1.2) monoclonal antibody (Product # 12-0903-82) in Flow Cytometry
Wed Feb 04 00:00:00 EST 2015
O-GlcNAcylation Negatively Regulates Cardiomyogenic Fate in Adult Mouse Cardiac Mesenchymal Stromal Cells.
PloS one
Zafir A,Bradley JA,Long BW,Muthusamy S,Li Q,Hill BG,Wysoczynski M,Prabhu SD,Bhatnagar A,Bolli R,Jones SP
12-0903 was used in Flow cytometry/Cell sorting to investigate how O-GlcNAcylation of intracellular proteins is involved in the regulation of mesenchymal stromal cell lineage commitment.
Wed Jun 29 00:00:00 EDT 2016
BMP-non-responsive Sca1+ CD73+ CD44+ mouse bone marrow derived osteoprogenitor cells respond to combination of VEGF and BMP-6 to display enhanced osteoblastic differentiation and ectopic bone formation.
PloS one
Madhu V,Li CJ,Dighe AS,Balian G,Cui Q
12-0903 was used in Flow cytometry/Cell sorting to study how combined action of VEGF and BMP on MSCs enhances osteoblastic differentiation of MSCs and increases their bone forming ability.
Fri Nov 20 00:00:00 EST 2015
Mouse Ly49G2+ NK cells dominate early responses during both immune reconstitution and activation independently of MHC.
Blood
Barao I,Alvarez M,Ames E,Orr MT,Stefanski HE,Blazar BR,Lanier LL,Anderson SK,Redelman D,Murphy WJ
12-0903 was used in Flow cytometry/Cell sorting to show that during reconstitution after HSCT and various activation stimuli, preferential expansion of the Ly49G2 receptor on NK cells occurs independently of MHC haplotype.
Thu Jun 30 00:00:00 EDT 2011
Hydrodynamic delivery of human IL-15 cDNA increases murine natural killer cell recovery after syngeneic bone marrow transplantation.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Barao I,Alvarez M,Redelman D,Weiss JM,Ortaldo JR,Wiltrout RH,Murphy WJ
12-0903 was used in Flow cytometry/Cell sorting to highlight cytokine gene transfer-based methods as a potential platform for natural killer cell recovery after bone marrow transplantation in mice.
Thu Dec 01 00:00:00 EST 2011
Characterization of fibroblasts responsible for cartilage destruction in arthritis.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Treese C,Mittag A,Lange F,Tarnok A,Loesche A,Emmrich F,Lehmann J,Sack U
12-0903 was used in Flow cytometry/Cell sorting to show that LS48 is an excellent tool to gain a deeper understanding of synovial fibroblasts in rheumatoid arthritis.
Tue Apr 01 00:00:00 EDT 2008
Hierarchy of Notch-Delta interactions promoting T cell lineage commitment and maturation.
The Journal of experimental medicine
Besseyrias V,Fiorini E,Strobl LJ,Zimber-Strobl U,Dumortier A,Koch U,Arcangeli ML,Ezine S,Macdonald HR,Radtke F
12-0903 was used in Flow cytometry/Cell sorting to study the Notch-Delta interactions that promote T cell development.
Mon Feb 19 00:00:00 EST 2007